1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carboxylic acid 97%
Supplier: Apollo Scientific
PC9463-1G PC9463-500MG
APOSPC9463-1GEA 666 EUR
APOSPC9463-1G APOSPC9463-500MG
1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carboxylic acid 97%
1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carboxylic acid
Formula: C₁₃H₈F₆N₂O₃
Melting Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD01936007
CAS Number: 158712-29-9