5-Chloro-2-fluorophenyl isothiocyanate 97%
Supplier: Apollo Scientific
Warning
PC9564-1G PC9564-5G PC9564-250MG
APOSPC9564-1GEA 72.7 EUR
APOSPC9564-1G APOSPC9564-5G APOSPC9564-250MG
5-Chloro-2-fluorophenyl isothiocyanate 97%
5-Chloro-2-fluorophenyl isothiocyanate
Formula: C₇H₃ClFNS
MW: 187.62 g/mol
Storage Temperature: Ambient
CAS Number: 247170-25-8