Tetrafluorobenzene-1,2-diol
Supplier: Apollo Scientific
Warning
PC9657-1G PC9657-250MG
APOSPC9657-1GEA 650 EUR
APOSPC9657-1G APOSPC9657-250MG
Tetrafluorobenzene-1,2-diol
Tetrafluorobenzene-1,2-diol
Formula: C₆H₂F₄O₂
Melting Pt: 68…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01075279
CAS Number: 1996-23-2