5-(Trifluoromethyl)-2-(4-trifluoromethylphenyl)-isoxazole-4-carboxylic acid 95+%
Supplier: Apollo Scientific
PC9720-1G PC9720-5G PC9720-25G
APOSPC9720-1GEA 229 EUR
APOSPC9720-1G APOSPC9720-5G APOSPC9720-25G
5-(Trifluoromethyl)-2-(4-trifluoromethylphenyl)-isoxazole-4-carboxylic acid 95+%
5-(Trifluoromethyl)-2-(4-trifluoromethylphenyl)-isoxazole-4-carboxylic acid
Formula: C₁₂H₇F₆NO₃
MW: 327.18 g/mol
Melting Pt: 180…185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD11100463
CAS Number: 886497-47-8