3,5-Difluoro-2-nitrophenol 98%
Supplier: Apollo Scientific
Warning
PC9743-1G PC9743-5G PC9743-10G
APOSPC9743-1GEA 29 EUR
APOSPC9743-1G APOSPC9743-5G APOSPC9743-10G
3,5-Difluoro-2-nitrophenol 98%
3,5-Difluoro-2-nitrophenol
Formula: C₆H₃F₂NO₃
MW: 175.0900 g/mol
Melting Pt: 46…47 °C
Storage Temperature: Ambient
MDL Number: MFCD08458165
CAS Number: 151414-46-9