Allyl-2,2,3,3,4,4,5,5-octafluoropentyl ether 97+%
Supplier: Apollo Scientific
Warning
PC9859-1G PC9859-5G
APOSPC9859-1GEA 79.9 EUR
APOSPC9859-1G APOSPC9859-5G
Allyl-2,2,3,3,4,4,5,5-octafluoropentyl ether 97+%
Allyl-2,2,3,3,4,4,5,5-octafluoropentyl ether
Formula: C₈H₈F₈O
Boiling Pt: 150…151 °C/749mmHg
Density: 1.392 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00443218
CAS Number: 3108-07-4