2-Fluoro-3-methylbenzaldehyde
Supplier: Apollo Scientific
Warning
PC9906-1G PC9906-5G
APOSPC9906-1GEA 58 EUR
APOSPC9906-1G APOSPC9906-5G
2-Fluoro-3-methylbenzaldehyde
2-Fluoro-3-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138.1390 g/mol
Boiling Pt: 77…79 °C/10mmHg
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD06797948
CAS Number: 886762-64-7