2-Fluoro-3-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Warning
PC9924-25G PC9924-5G PC9924-100G
APOSPC9924-25GEA 79.9 EUR
APOSPC9924-25G APOSPC9924-5G APOSPC9924-100G
2-Fluoro-3-(trifluoromethyl)pyridine 98%
2-Fluoro-3-(trifluoromethyl)pyridine
Formula: C₆H₃F₄N
MW: 165.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973416
CAS Number: 65753-52-8