1-Acetyl-4-fluoronaphthalene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4'-Fluoro-1'-acetonaphthone, 1-(4-Fluoronaphthalen-1-yl)ethanone, 1-(4-Fluoro-1-naphthyl)ethanone
PC9950-1G PC9950-25G PC9950-5G
APOSPC9950-1GEA 21.9 EUR
APOSPC9950-1G APOSPC9950-25G APOSPC9950-5G
1-Acetyl-4-fluoronaphthalene 97%
1-Acetyl-4-fluoronaphthalene
Formula: C₁₂H₉FO
Boiling Pt: 164…166 °C/14mmHg
Melting Pt: 33…35 °C
Density: 1.203 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134475
CAS Number: 316-68-7
EINECS: 206-261-9