3-Fluoro-4-nitrobenzonitrile 99%
Supplier: Apollo Scientific
Warning
PC9951-25G PC9951-100G PC9951-10G
APOSPC9951-25GEA 94.4 EUR
APOSPC9951-25G APOSPC9951-100G APOSPC9951-10G
3-Fluoro-4-nitrobenzonitrile 99%
3-Fluoro-4-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD04973417
CAS Number: 218632-01-0