Aprotinin
Supplier: Apollo Scientific
BIMI2132-100MG BIMI2132-250MG
APOSBIMI2132-100MGEA 242 EUR
APOSBIMI2132-100MG APOSBIMI2132-250MG
Aprotinin
Proteins and Peptides
Aprotinin inhibits trypsin, chymotrypsin, kallikrein from different sources and plasmin.