Universal reference RNAs
Supplier: AGILENT
740000 740100 740200
AGLS740000EA 988 EUR
AGLS740000 AGLS740100 AGLS740200
Universal reference RNAs
Nucleic Acid Reagents RNAi
High-quality total RNA for human, mouse, and rat microarray gene-expression profiling that acts as a consistent control for standard data set comparisons.
  • Comprised of 10 cell lines for human, 11 for mouse and 14 for rat giving broad gene coverage
  • UHRR selected as one of the two standards in the FDA led Microarray Quality Control (MAQC) project
The reference RNA is designed to be used as a reference for uarray gene-profiling experiments.
Delivery information: Includes RNase-free water, 1,5 ml.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more