GC columns, J&W Ultra 1
Supplier: Agilent
19091A-002 19091A-005 19091A-012 19091A-015 19091A-101 19091A-102 19091A-105 19091A-112 19091A-115 19091A-102E 19091A-008 19091A-108
HEWL19091A-002EA 1080 EUR
HEWL19091A-002 HEWL19091A-005 HEWL19091A-012 HEWL19091A-015 HEWL19091A-101 HEWL19091A-102 HEWL19091A-105 HEWL19091A-112 HEWL19091A-115 HEWL19091A-102E HEWL19091A-008 HEWL19091A-108
GC columns, J&W Ultra 1
Chromatography Columns
Agilent's non-polar Ultra 1 columns deliver excellent performance and accuracy in a range of chromatographic applications.
  • 100% Dimethylpolysiloxane
  • Non-polar
  • Equivalent to HP-1 with tighter specifications for retention index and capacity factors
  • Bonded and cross-linked
  • Solvent rinsable
This is a nonpolar column that resembles HP-1 but with tighter specifications for retention index and capacity factors. The column has an operating temperature range of –60 to +325 °C/350 °C and is intended for use with the Sadtler Retention Index Library. Precision-engineered Ultra 1 columns are bonded, cross-linked and solvent rinsable, and are delivered tested to some of the strictest industry QC standards, ensuring confidence in the accuracy and integrity of analytical results every time.