Scrubber adaptors for bottles with GL thread, BOLA
Supplier: Bohlender
N1660-14 N1660-24
201-0284EA 141 EUR
201-0284 201-0285
Scrubber adaptors for bottles with GL thread, BOLA
Bottles Gas Washing Bottles
Screw cap made from PP, base body made from PTFE and gas inlet pipe/gas distributor made from FEP.
  • Excellent good chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −200 to +250 °C
  • Suitable for vacuum applications
Resistance-free gas inlet and discharge pipes with hard-walled hoses (e.g. PTFE), which are connected via optionally available laboratory screw connections. For elastic hoses, please use GL hose connections. The inlet tube can be shortened to the desired length. The retainer screw cap can beMore Product Information