General purpose gloves, Perfect Poly® Workeasy Black black
Supplier: HONEYWELL SAFETY
2100251/10 2100251/8 2100251/7 2100251/9
112-1631EA 34.1 EUR
112-1631 112-1633 112-1632 112-1634
General purpose gloves, Perfect Poly® Workeasy Black black
Gloves
Knitted glove made of 100% black polyester with black polyurethane coating on palm and fingertips. The polyester and its polyurethane coating provide good abrasion and tear resistance.
  • Tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • PU coating ensures a good grip
  • Ventilated back and PU coating optimise the glove's breathability
  • Certified as 'silicone-free' to ensure their use in painting
  • Its dark colour makes it more soilproof, thus ensuring a longer life
Ideal for the handling of fine products in a dry, slightly greasy or soiled environment.
Gauge: 13
Colour: Black
EN 388 (3121); Cat. II
Certifications: EN 388:2016 (2121X); Cat. II.