Hand disinfectant, Desderman® Care
Supplier: SCHUELKE MAYR
70002056 70002343
SCUE70002056EA 205 EUR
SCUE70002056 SCUE70002343
Hand disinfectant, Desderman® Care
Skincare Skin Disinfectants Gel Skin Disinfectants
Alcohol-based hand rub for hygienic and surgical hand disinfection with panthenol and vitamin E, without colour and perfume.
  • Broad spectrum of efficacy (incl. viruses according EN 14476)
  • Outstanding skin feeling through an innovative skin care formula (ProPanthenol-complex and vitamin E)
  • Care effect of the skin care formular clinically confirmed
  • Biocidal product
Hygienic hand disinfection (EN 13727 + EN 1500)
Surgical hand disinfection (EN 13727 + EN 12791)
The product is to be used undiluted. During the contact time for hygienic and surgical hand disinfection, the product should be applied in portions so that the hands or hands and forearms remain well moistened during the entire application time.
Products are manufactured economically and with advanced, safe and environmentally friendly production processes while at the same time maintaining high quality standards.
100 g solution contains the following active ingredients: 89,1 g Ethanol 96% (v/v)
Form: liquid
Density: approx. 0,83 g/cm3 / 20 °C
Flash point: 17 °C / Method: DIN EN ISO 13736
Color: colourless
Certifications: IHO listed
ÖGHMP certificate
VAH certificate
Caution: Use disinfectants safely. Always read the label and product information before use. For external application. Do not bring into contact with open flames or electrical heat sources that are switched on.