Celite® 503, filter aid, GPR RECTAPUR®
Supplier: VWR Chemicals
22553.290EA 35.3 EUR
22553.290 22553.360
Celite® 503, filter aid, GPR RECTAPUR®
Celite® 503, filter aid
Celite® is used as a filtering aid or as adsorbent in column chromatography.
  • Filter aid
  • Flux Calcined Diatomite
Formula: O₂Si
MW: 60.08 g/mol
CAS Number: 68855-54-9
Order Now
Specification Test Results
Identification Passes test
pH (25 °C; 10%) 8,5 - 11,0
Wet density ≤ 369 g/l
Loss on ignition (600 °C) ≤ 0.5%
Particle size (< 0.1 mm) ≥ 85%