Tubing rolls, PTFE, BOLA
Supplier: Bohlender
S2010-41 S2010-42 S2010-50 S2010-60 S2010-64 S2010-65 S2012-10 S2012-26 S2012-30 S2012-32 S2012-40
BOHLS2010-41EA 135 EUR
BOHLS2010-41 BOHLS2010-42 BOHLS2010-50 BOHLS2010-60 BOHLS2010-64 BOHLS2010-65 BOHLS2012-10 BOHLS2012-26 BOHLS2012-30 BOHLS2012-32 BOHLS2012-40
Tubing rolls, PTFE, BOLA
Tubing Transfer Tubing
Translucent to milky-white appearance.
  • Almost universal chemical resistance
  • Free from extractable agents
  • Physiologically safe
  • Non-adhesive surface
  • Very good sliding and dielectric characteristics
  • Oxygen value more than 95
  • Resistant to irradiation and weather
Perfect tubing for aggressive and pure liquids or gases. Can be sterilized in ETO and in autoclaves.
Certifications: Flame retardant according to UL94V0.