Heating and drying ovens, Universal UF/UFplus (forced convection)
Supplier: MEMMERT
UF55 UF55PLUS UF75 UF750 UF750PLUS UF75PLUS UF110 UF110PLUS UF160 UF160PLUS UF260 UF260PLUS UF30 UF30PLUS UF450 UF450PLUS UF1060 UF1060PLUS UF1060MPLUS UF110M UF110MPLUS UF55M UF160M UF55MPLUS UF160MPLUS UF75M UF260M UF75MPLUS UF260MPLUS UF750M UF30M UF750MPLUS UF30MPLUS UF450M UF450MPLUS UF1060M
466-0355EA 1763 EUR
466-0355 466-0356 466-0357 466-0358 466-0359 466-0360 466-0345 466-0346 466-0347 466-0348 466-0349 466-0350 466-0351 466-0352 466-0353 466-0354 466-0433 466-0434 MEMMUF1060MPLUS MEMMUF110M MEMMUF110MPLUS MEMMUF55M MEMMUF160M MEMMUF55MPLUS MEMMUF160MPLUS MEMMUF75M MEMMUF260M MEMMUF75MPLUS MEMMUF260MPLUS MEMMUF750M MEMMUF30M MEMMUF750MPLUS MEMMUF30MPLUS MEMMUF450M MEMMUF450MPLUS MEMMUF1060M
Heating and drying ovens, Universal UF/UFplus (forced convection)
Ovens Standard Ovens
The range includes forced convection models with a temperature range up to 300 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application.
  • SingleDISPLAY models, ideal for standard applications:
  • ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, program time, time zones and daylight saving time
  • One PT100 temperature sensor in a four wire circuit and Ethernet interface for reading out the protocol log
  • Double over-temperature protection including visual alarm: Electronic temperature monitoring with freely adjustable monitoring temperature and mechanical temperature limiter TB acc. to DIN 12880 which switches off the heating if the maximum permitted temperature is exceeded by approx. 10 °C, protecting the oven
All models have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. These ovens have an integrated data logger and digital timer, which is adjustable from 1 minute to 99 days, 23 hours. The SetpointWAIT function ensures the process time does not start before the set temperature is reached. This feature can also be used with additional, freely positionable PT100 sensors, so that the process time starts when the set temperature is reached at all measuring points. All models with TwinDISPLAY feature the intuitive control and logging software, AtmoCONTROL, which uses drag and drop symbols to input values. The software allows online monitoring of up to 32 connected appliances and can send an automatic alarm message to one or several email addresses. Complex ramping processes can be programmed quickly and easily for TwinDISPLAY models.
TwinDISPLAY models, ideal for complex ramping processes:
Intuitive, user friendly ovens with 2 TFT displays. ControlCOCKPIT with USB port. ControlCOCKPIT parameters as above
Two PT100 sensors in a 4-wire circuit for mutual monitoring, taking over functions in case of an error
The HeatBALANCE function allows application specific adjustment of heating power distribution (balance) between the upper and lower heating areas
Logged protocol data is displayed on the ControlCOCKPIT (max. 10000 values correspond to approx. 1 week)
Ethernet interface for reading out the protocol log and for uploading and implementing programs and for online logging
Multiple over-temperature protection with audible and visual alarm: Electronic temperature monitoring TWW/TWB adjustable on display (independent adjustable temperature safety device Class 3.1, which protects the samples and the oven) and mechanical temperature limiter TB (protection Class 1) acc. to. DIN 12880. AutoSAFETY automatically adjusts to the set value within a freely adjustable tolerance range. Setting individual MIN/MAX values for over-/under-temperature
The Um ovens, which have the same specifications as standard models, are classified as a Class I medical device in accordance with the EU directive 93/42/EEC.
Possible applications as a medical device:
The UFm or UFmplus models, equipped with a fan for forced air circulation, can be used for warming non-sterile blankets and cloths.
Certifications: IP protection class according to DIN EN 60529: IP 20
Accessories information: For details of the range of shelves and grids, please visit website or contact your local Avantor sales office.
Ordering information: Supplied without shelves, please order required shelves, stainless steel grids or trays separately. For all chamber volumes >53 litres, a minimum of 2 shelves or grids must be ordered in addition. Range of accessories and factory fitted options such as access ports, interior sockets, lighting and interior door are available on request. Please contact your local Avantor sales office for further details prior to ordering as these cannot be retro-fitted. Please note U 450 and U 750 units require a three phase supply.