Cryogenic Direct Thermal Labels
Supplier: GA
DFP-103C1-2WH DFP-103C3-2WH DFP-10C1-1WH DFP-10C3-2WH DFP-116C1-6WH DFP-116C3-6WH DFP-127C1-1WH DFP-127C3-1WH DFP-129C1-1WH DFP-129C3-2WH DFP-150C1-1WH DFP-150C3-3WH DFP-161C1-2WH DFP-161C3-2WH DFP-177C1-1WH DFP-177C3-3WH DFP-18C1-2WH DFP-18C3-2WH DFP-204C1-2WH DFP-204C3-2WH DFP218C1-0.5WH DFP-218C3-1WH DFP-227C1-2WH DFP-227C3-4WH DFP-245C1-1WH DFP-245C3-3WH DFP-24C1-2WH DFP-24C3-2WH DFP-257C1-3WH DFP257C3-7.5WH DFP-25C1-1WH DFP-25C3-2WH DFP-271C1-3WH DFP-271C3-6WH DFP-28C1-1WH DFP-28C3-2WH DFP-29C1-1WH DFP-29C3-1WH DFP-32C1-1WH DFP-32C3-3WH DFP-4C1-1WH DFP-4C3-5WH DFP-504C1-1WH DFP-504C3-3WH DFP52NOTC1-2WH DFP52NOTC3-2WH DFP-56C1-1WH DFP-56C3-1WH DFP-57C1-2WH DFP-57C3-2WH DFP-5C1-1WH DFP-5C3-2WH DFP61NPNOT14WH DFP61NPNOT34WH DFP-6C1-1WH DFP-6C3-3WH DFP-70C1-2WH DFP-70C3-2WH DFP-75NPC1-2WH DFP-75NPC3-2WH DFP-7C1-1WH DFP-7C3-1WH DFP-94C1-1WH DFP-94C3-1WH DFP-95C1-8WH DFP-95C3-8WH DFP-97C1-1WH DFP-97C3-1WH DFP-9C1-1WH DFP-9C3-1WH DFPC-129C1-1BL DFPC-129C1-1GA DFPC-129C1-1LA DFPC-129C1-1MT DFPC-129C1-1OR DFPC-129C1-1PI DFPC-129C1-1YE DFPC-129C3-2BL DFPC-129C3-2GA DFPC-129C3-2LA DFPC-129C3-2MT DFPC-129C3-2OR DFPC-129C3-2PI DFPC-129C3-2YE DFPC-161C1-2BL DFPC-161C1-2LE DFPC-161C1-2OR DFPC-161C3-2BL DFPC-161C3-2LE DFPC-161C3-2OR DFPC-202C3-1BL DFPC218C10.5BL DFPC-218C3-1BL DFPC-28C1-1BL DFPC-28C1-1GA DFPC-28C3-2BL DFPC-28C3-2GA
GAINDFP-103C1-2WHEA 123 EUR
GAINDFP-103C1-2WH GAINDFP-103C3-2WH GAINDFP-10C1-1WH GAINDFP-10C3-2WH GAINDFP-116C1-6WH GAINDFP-116C3-6WH GAINDFP-127C1-1WH GAINDFP-127C3-1WH GAINDFP-129C1-1WH GAINDFP-129C3-2WH GAINDFP-150C1-1WH GAINDFP-150C3-3WH GAINDFP-161C1-2WH GAINDFP-161C3-2WH GAINDFP-177C1-1WH GAINDFP-177C3-3WH GAINDFP-18C1-2WH GAINDFP-18C3-2WH GAINDFP-204C1-2WH GAINDFP-204C3-2WH GAINDFP218C1-0.5WH GAINDFP-218C3-1WH GAINDFP-227C1-2WH GAINDFP-227C3-4WH GAINDFP-245C1-1WH GAINDFP-245C3-3WH GAINDFP-24C1-2WH GAINDFP-24C3-2WH GAINDFP-257C1-3WH GAINDFP257C3-7.5WH GAINDFP-25C1-1WH GAINDFP-25C3-2WH GAINDFP-271C1-3WH GAINDFP-271C3-6WH GAINDFP-28C1-1WH GAINDFP-28C3-2WH GAINDFP-29C1-1WH GAINDFP-29C3-1WH GAINDFP-32C1-1WH GAINDFP-32C3-3WH GAINDFP-4C1-1WH GAINDFP-4C3-5WH GAINDFP-504C1-1WH GAINDFP-504C3-3WH GAINDFP52NOTC1-2WH GAINDFP52NOTC3-2WH GAINDFP-56C1-1WH GAINDFP-56C3-1WH GAINDFP-57C1-2WH GAINDFP-57C3-2WH GAINDFP-5C1-1WH GAINDFP-5C3-2WH GAINDFP61NPNOT14WH GAINDFP61NPNOT34WH GAINDFP-6C1-1WH GAINDFP-6C3-3WH GAINDFP-70C1-2WH GAINDFP-70C3-2WH GAINDFP-75NPC1-2WH GAINDFP-75NPC3-2WH GAINDFP-7C1-1WH GAINDFP-7C3-1WH GAINDFP-94C1-1WH GAINDFP-94C3-1WH GAINDFP-95C1-8WH GAINDFP-95C3-8WH GAINDFP-97C1-1WH GAINDFP-97C3-1WH GAINDFP-9C1-1WH GAINDFP-9C3-1WH GAINDFPC-129C1-1BL GAINDFPC-129C1-1GA GAINDFPC-129C1-1LA GAINDFPC-129C1-1MT GAINDFPC-129C1-1OR GAINDFPC-129C1-1PI GAINDFPC-129C1-1YE GAINDFPC-129C3-2BL GAINDFPC-129C3-2GA GAINDFPC-129C3-2LA GAINDFPC-129C3-2MT GAINDFPC-129C3-2OR GAINDFPC-129C3-2PI GAINDFPC-129C3-2YE GAINDFPC-161C1-2BL GAINDFPC-161C1-2LE GAINDFPC-161C1-2OR GAINDFPC-161C3-2BL GAINDFPC-161C3-2LE GAINDFPC-161C3-2OR GAINDFPC-202C3-1BL GAINDFPC218C10.5BL GAINDFPC-218C3-1BL GAINDFPC-28C1-1BL GAINDFPC-28C1-1GA GAINDFPC-28C3-2BL GAINDFPC-28C3-2GA
Cryogenic Direct Thermal Labels
Labels
These thermoplastic waterproof labels are specifically designed to permanently adhere to vials, microcentrifuge tubes, microtubes, microplates, glass tubes, freezer boxes, plastic bags, and other specimen containers that will undergo long-term cryogenic and deep-freeze storage.
  • Temperature range: –196 to +93 °C (–321 to +199 °F)
  • Labels can withstand long-term storage in liquid nitrogen (–196° C), laboratory freezers (–80 °C, –20 °C), and transportation in dry ice
  • Ideal for the identification of vials, microtubes, microplates, and freezer boxes
  • Printable with all major direct thermal printer brands
  • No need for extra lamination
  • Strong permanent adhesive ensures labels will not wrinkle or peel
Providing a strong affinity to glass, polypropylene, polyethylene, and other plastic surfaces, these barcode labels will retain their adhesion to tubes through multiple freeze-thaw cycles without cracking or peeling off. Their slim design ensures these direct thermal cryogenic labels are compatible with most cryogenic storage systems, with minimal risk of being scraped off.