4N6FLOQSwabs® Forensic DNA Collection for Human Identification
Supplier: Copan
4520CS01 4508C 4504C 4520CA 40U022D
COPA4520CS01EA 1160 EUR
COPA4520CS01 COPA4508C COPA4504C COPA4520CA COPA40U022D
4N6FLOQSwabs® Forensic DNA Collection for Human Identification
Swabs Swabs for Sampling
DNA collection for human identification. The active drying system absorbs water molecules, preventing microbial growth and guaranteeing DNA stability for 12+ months at RT and –80 °C.
  • Application: High performance sample collection 
  • FLOQ® technology: this Swab is empowered by the FLOQ® technology for superior sample adsorption and release
  • 20 mm breaking point: the short length breaking point and the 2-1,5 ml vials compatible tip facilitate the laboratory workflow
  • Gold standard for forensic DNA evidence collection: 4N6FLOQSwabs® w/Active drying system have been extensively adopted by law enforcement agencies for the evidence collection at a crime scene
  • Different shapes for different collection: 4N6FLOQSwabs® are available with different geometries to ensure that the maximum amount of sample can be collected also from the most challenging surfaces
  • Forensic DNA grade: ISO 18385:2016 compliance minimizes the risk of detectable human nuclear DNA contamination in products used by the global forensic community
For more than ten years, our 4N6 FLOQSwabs® Genetics line led the development of a new quality standard for forensics applications. Thanks to our partnership with key opinion leaders and our strong know-how, we delivered a first-rate product line for the whole range of forensic applications such as: databasing, sexual assault, paternity testing, evidence collection, genotyping, and skin microbiome.
The validated EO TREATMENT assesses the degradation of possible human DNA residuals.
- Free of amplifiable human DNA, detectable and amplifiable DNase, and RNase.
- personnel STR database
- stringent controls and automated manufacturing
processes.
Certifications:
ISO 18385:2016 compliant.
Caution: 4N6FLQ Swabs® Genetics are not intended for diagnostic purposes.