Gum tragacanth, GPR RECTAPUR®

Supplier: VWR Chemicals

84110.290EA 0 EUR
84110.290 84110.460 84110.260
Gum tragacanth, GPR RECTAPUR®
Gum tragacanth
Storage Temperature: Ambient
MDL Number: MFCD00131255
CAS Number: 9000-65-1
EINECS: 232-552-5

Order Now

Specification Test Results

Identification Passes test
Heavy metals (as Pb) ≤ 20 ppm
Loss on drying (105 °C) ≤ 15 %
Total ash ≤ 4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR