Tubes made with 90% biobased plastic. To be traced back to biological waste material. Available with screw cap in volumes 5,0 to 50 ml. Ideal for cell culture, cell biology applications, sample preparation in microbiology and molecular biology.
- Premium biobased PP - produced from recycled feedstock in second generation - in this case derived solely from waste and residue streams from e.g., vegetable oil production for instance used cooking oil
- Premium USP class 6 raw material and no use of slip agents, plasticizers, and biocides during manufacturing ensure highest sample integrity
These tubes in the sizes 5,0, 15, 25 and 50 ml open new doors to make laboratory work significantly more sustainable without jeopardizing experimental results. By using a second-generation bio circular polymer made from 90% renewable resources (e.g., waste and residues from vegetable oil refining or waste cooking oil) the product-related carbon footprint of these tubes was significantly reduced.
'General Quality Certificates' on compliance with standards and general quality items, product specific and lot specific purity certificates as well as 'ACT Certificates' (the independent environmental impact factor certification by My Green Lab) certify the well-known premium product quality you can expect from Eppendorf Tubes combined with a sustainability factor that will clearly be visible with continuous use of these tubes in your laboratory carbon footprint.
Like the tubes made of virgin PP also these tubes are your solution for cell culture and cell biology applications as well as for sample preparation protocols in microbiology and molecular biology laboratories. Whether centrifugation, mixing, or other applications, the Eppendorf Tubes BioBased in 5,0, 15, 25 and 50 ml – with screw cap – will complete your workflows in an excellent way. Additionally, they support you in your efforts to become visibly more environmentally friendly in the laboratory as well.
Certifications: ACT (Accountability, Consistency, and Transparency) Certification - the independent validation provided by 'My Green Lab'. ISCC PLUS (International Sustainability and Carbon Certification) to enable traceability along the entire supply chain. Independent laboratory - Lot specific purity certificates can be downloaded from Eppendorf website. General Quality Certificates on compliance with standards and general Eppendorf quality items.