Pomalidomide
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-Amino-N-(2,6-dioxo-3-piperidyl)phthalimide, 4-Amino-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione
465700010 465702500
ACRO465700010EA 328 EUR
ACRO465700010 ACRO465702500
Pomalidomide
Pomalidomide
Formula: C₁₃H₁₁N₃O₄
MW: 273.25 g/mol
Melting Pt: 315°C…320°C
Storage Temperature: Refrigerator
MDL Number: MFCD12756407
CAS Number: 19171-19-8
Order Now
Specification Test Results
Appearance Yellow to green powder
Infrared spectrum Conforms
HPLC ≥ 98.5%
Loss on drying ≤ 1%