Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) Pd 12,1%, crystalline solid
Supplier: Thermo Fisher Scientific
Warning
Synonyms: [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]palladium(II) Dichloride, [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]dichloropalladium(II)
46665.03 46665.06
46665.03EA 146 EUR
46665.03 46665.06
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) Pd 12,1%, crystalline solid
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)
Formula: C₃₉H₃₂Cl₂OP₂Pd
MW: 755.95 g/mol
Melting Pt: 280…287 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD14155707
CAS Number: 205319-10-4