Indigo carmine
Supplier: Spectrum Chemical
Warning
Synonyms: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue
FD115-100GM FD115-25GM FD115-500GM
SPCMFD115-100GMEA 184 EUR
SPCMFD115-100GM SPCMFD115-25GM SPCMFD115-500GM
Indigo carmine
Indigo carmine
FD and C Blue No. 2 - In the United States, the Food and Drug Administration (FDA) controls the specifications for Food, Drug and Cosmetic (FD&C) colours.
Spectrum certifies all batches of FD&C colours meet the FDA's quality standards.
Caution: Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.
Formula: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005723
CAS Number: 860-22-0
Merck Index: 13,04969
Order Now
Specification Test Results
Identification To Pass Test