ESD safety ankle boots, lace-up, Flash 6805 XP
Supplier: ATLAS - the shoe company
75800/36 75800/37 75800/38 75800/39 75800/40 75800/41 75800/42 75800/43 75800/44 75800/45 75800/46 75800/47 75800/48 75800/49
114-9228EA 169 EUR
114-9228 114-9229 114-9230 114-9231 114-9232 114-9233 114-9234 114-9235 114-9236 114-9237 114-9238 114-9239 114-9240 114-9241
ESD safety ankle boots, lace-up, Flash 6805 XP
Shoes and Boots
ESD safety ankle boot made from a water-repellent, quick-drying and durable texile upper (Sportline) and a MPU® INNOFLEX sole.
  • Clima-stream® technology ensures moisture regulation and breathability
  • Skin-friendly, abrasion-resistant active X lining for an improved climate
  • 3D cushioning system: Pressure-relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • alu-tec® aluminium toecap (200 J)
  • XP® metal free puncture-resistance
  • Energy absorbing MPU® INNOFLEX sole technology reduces fatigue
  • Removable insoles; orthopedic insoles possible
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Colour: Black
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3 SRC