Accessories for Monitor UPC-900
Supplier: Cytiva
18-1128-24 18-1128-95 18-0620-01 18-0621-01 18-0622-01 18-0623-01 18-0624-01 18-0625-01 18-0626-01 18-0627-01 18-1111-14
18-1128-24EA 2047 EUR
18-1128-24 18-1128-95 18-0620-01 18-0621-01 18-0622-01 18-0623-01 18-0624-01 18-0625-01 18-0626-01 18-0627-01 18-1111-14
Accessories for Monitor UPC-900
Electrochemistry Meters pH and Multi-Parameter Meters
Order Now
SPECIFICATIONS
Model Monitor UPC-900
Environmentally Preferable
pH range 0 - 14 (specifications valid from 2 - 12)
pH accuracy ± 0,1 pH unit, temperature compensated
Conductivity range 1 μS/cm - 999,9 mS/cm
Outputs Analog, 0 - 1 V full-scale
W×D×H 260×370×100 mm