Pharmaceutical Perfection refrigerators compliant with DIN 13277, HMFvh
Supplier: Liebherr
HMFVH4001 HMFVH4001H63 HMFVH4011 HMFVH4011H63 HMFVH5501 HMFVH5501H63 HMFVH5511 HMFVH5511H63
LIEBHMFVH4001EA 2979 EUR
LIEBHMFVH4001 LIEBHMFVH4001H63 LIEBHMFVH4011 LIEBHMFVH4011H63 LIEBHMFVH5501 LIEBHMFVH5501H63 LIEBHMFVH5511 LIEBHMFVH5511H63
Pharmaceutical Perfection refrigerators compliant with DIN 13277, HMFvh
Refrigerators
Ultra-efficient pharmaceutical refrigerators have been designed for the particular requirements with their dynamic cooling system, highly effective insulation, and precise temperature control, they guarantee reliable storage of sensitive substances and preparations.
  • Integrated data logger and USB interface for reading the data logger
  • PT1000 product sensor and Sensor feedthrough
  • Min./max. temperature log shows whether the storage temperatures were within the tolerance limits
  • Insulated and glass door models with vertical lighting, self-closing door at <90°
More Product Information
Order Now
SPECIFICATIONS
Model HMFvh 4001 HMFvh 4001 H63 HMFvh 4011 HMFvh 4011 H63 HMFvh 5501 HMFvh 5501 H63 HMFvh 5511 HMFvh 5511 H63
Environmentally Preferable
Display 2,4" touch and swipe colour display 2,4" touch and swipe colour display 2,4" touch and swipe colour display 2,4" touch and swipe colour display 2,4" touch and swipe colour display 2,4" touch and swipe colour display 2,4" touch and swipe colour display 2,4" touch and swipe colour display
Defrost Automatic Automatic Automatic Automatic Automatic Automatic Automatic Automatic
Alarm Visual and audible Visual and audible Visual and audible Visual and audible Visual and audible Visual and audible Visual and audible Visual and audible
Access ports 1×Ø 10 mm 1×Ø 10 mm 1×Ø 10 mm 1×Ø 10 mm
Shelves/drawers 6 (5 adjustable), wire shelves plastic-coated 1 adjustable, wire shelf plastic-coated/8 drawers 6 (5 adjustable), wire shelves plastic-coated 1 adjustable, wire shelf plastic-coated/8 drawers 6 (5 adjustable), wire shelves plastic-coated 1 adjustable, wire shelf plastic-coated/7 drawers 6 (5 adjustable), wire shelves plastic-coated 1 adjustable, wire shelf plastic-coated/7 drawers
Door type steel steel glass glass steel steel glass glass
Door lock electronic, with remote control electronic, with remote control electronic, with remote control electronic, with remote control electronic, with remote control electronic, with remote control electronic, with remote control electronic, with remote control
Refrigerant R 600a R 600a R 600a R 600a R 600a R 600a R 600a R 600a
Lamp LED lighting column, left Vertical LED light strip recessed in left wall with separate switch Vertical LED light strip recessed in left wall with separate switch Vertical LED light strip recessed in left wall with separate switch LED lighting column, left Vertical LED light strip recessed in left wall with separate switch Vertical LED light strip recessed in left wall with separate switch Vertical LED light strip recessed in left wall with separate switch
Sound level 49 dB(A) 49 dB(A) 49 dB(A) 49 dB(A) 49 dB(A) 49 dB(A) 49 dB(A) 49 dB(A)
Electrical 220 - 240V~, 2,0 A 220 - 240V~, 2,0 A 220 - 240V~, 2,0 A 220 - 240V~, 2,0 A 220 - 240V~, 2,0 A 220 - 240V~, 2,0 A 220 - 240V~, 2,0 A 220 - 240V~, 2,0 A
Weight 73 (gross)/64,5 (net) kg 69 (gross)/64,5 (net) kg 89 (gross)/83 (net) kg 82 (gross)/69 (net) kg 91 (gross)/82 (net) kg 106 (gross)/100 (net) kg 111 (gross)/102 (net) kg 101 (gross)/97 (net) kg