Cleanroom gloves, polyisoprene, sterile, Biogel® PI Micro Tech
Supplier: MOLNLYCKE HEALTH CARE
4448555 4448560 4448565 4448570 4448575 4448580 4448585 4448590
112-6378EA 582 EUR
112-6378 112-6379 112-6380 112-6381 112-6382 112-6383 112-6384 112-6385
Cleanroom gloves, polyisoprene, sterile, Biogel® PI Micro Tech
Gloves
These sterile, cleanroom gloves with beaded anti-slip cuffs and curved fingers are suitable for use in aseptic and Class 100 (ISO 5)/EU GMP Grade A environments.
  • Can be worn together with Biogel® PI Micro Tech Indicator® underglove to form a Breach Indication System
  • Exceed industry standard for force at break, tensile strength and elongation
  • Tested and cleared for use with chemotherapy agents
  • Synthetic polyisoprene
  • Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Ideal for use in controlled environments, in medical and research laboratories, in the pharma industry, as well as in biotechnology.
Certifications: EN 455, ASTM D3577, ISO 10282, EN 374, EN 16523-1, PPE Cat. III, AQL 0,65, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, ASTM F1671, Allergenicity ISO 10993, Pyrogenicity ASTM D7102, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420.
Packaging: One pair per pack, in a high quality polyethylene inner wrap, packed into a film pack. 25 pairs per inner LDPE polybag f; 2 inner LDPE polybags are packed in an outer LDPE polybag. Four outer polybags per transit case, total of 200 pairs.