Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™
Supplier: Thermo Fisher Scientific
Quanti-Cult Plus™
OXOIR4718001EA 1740 EUR
OXOIR4718001
Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™
Microbiology Test Systems Microorganism Tests
Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™ is supplied as a film of preserved microorganisms inside the cap on a plastic vial.
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.