PermaSleeve® Heat-Shrink Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Supplier: Brady
173615 173680 173631 173673 173661 173645 173590 173584 173603 173641 173654 173980 173644 173974 173610 173674 173655 173979 173597 173591 173643 173608 173638 173607 173642 173668 173663 173676 173650 173627 173622 173630 173665 173647 173612 173972 173596 173592 173659 173637 173582 173595 173632 173640 173648 173652 173678 173580 173588 173981 173623 173976 173639 173583 173636 173649 173589 173977 173982 173593 173666 173681 173620 173601 173660 173971 173670 173614 173613 173617 173618 173651 173598 173978 173653 173662 173585 173604 173606 173616 173669 173626 173611 173646 173602 173609 173633 173600 173619 173975 173675 173587 173594 173671 173667 173677 173664 173657 173586 173634 173599 173625 173629 173621 173658 173624 173679 173605 173973 173682
BRDY173615EA 113.28 EUR
BRDY173615 BRDY173680 BRDY173631 BRDY173673 BRDY173661 BRDY173645 BRDY173590 BRDY173584 BRDY173603 BRDY173641 BRDY173654 BRDY173980 BRDY173644 BRDY173974 BRDY173610 BRDY173674 BRDY173655 BRDY173979 BRDY173597 BRDY173591 BRDY173643 BRDY173608 BRDY173638 BRDY173607 BRDY173642 BRDY173668 BRDY173663 BRDY173676 BRDY173650 BRDY173627 BRDY173622 BRDY173630 BRDY173665 BRDY173647 BRDY173612 BRDY173972 BRDY173596 BRDY173592 BRDY173659 BRDY173637 BRDY173582 BRDY173595 BRDY173632 BRDY173640 BRDY173648 BRDY173652 BRDY173678 BRDY173580 BRDY173588 BRDY173981 BRDY173623 BRDY173976 BRDY173639 BRDY173583 BRDY173636 BRDY173649 BRDY173589 BRDY173977 BRDY173982 BRDY173593 BRDY173666 BRDY173681 BRDY173620 BRDY173601 BRDY173660 BRDY173971 BRDY173670 BRDY173614 BRDY173613 BRDY173617 BRDY173618 BRDY173651 BRDY173598 BRDY173978 BRDY173653 BRDY173662 BRDY173585 BRDY173604 BRDY173606 BRDY173616 BRDY173669 BRDY173626 BRDY173611 BRDY173646 BRDY173602 BRDY173609 BRDY173633 BRDY173600 BRDY173619 BRDY173975 BRDY173675 BRDY173587 BRDY173594 BRDY173671 BRDY173667 BRDY173677 BRDY173664 BRDY173657 BRDY173586 BRDY173634 BRDY173599 BRDY173625 BRDY173629 BRDY173621 BRDY173658 BRDY173624 BRDY173679 BRDY173605 BRDY173973 BRDY173682
PermaSleeve® Heat-Shrink Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Labels
PermaSleeve® PS heat-shrink polyolefin (B-342) wire marking sleeves combine speed, ease of use, and permanence to create long-lasting, high-quality identification.
  • 3:1 heat shrinkable sleeve fit snugly around wires for maximum insulation protection and permanent identification
  • Fade-resistant and smudge-proof for superior aesthetic appearance
  • Cold resistant and flame-retardant with an operating temperature of –55 to +135 °C
Applications include electrical labelling and wire and cable labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/Bradyprinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certifications: Meets SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202. RoHS compliant. UL recognised.