Hi-Vis Flame Resistant Vests
Supplier: Portwest
FR75YER4X/5X FR75YERL/XL FR75YERS/M FR75YERXX/3X
PRTWFR75YER4X/5XEA 20.3 EUR
PRTWFR75YER4X/5X PRTWFR75YERL/XL PRTWFR75YERS/M PRTWFR75YERXX/3X
Hi-Vis Flame Resistant Vests
Workwear
The FR75 is a great addition to our flame retardant vest collection.
  • Premium sewn-on flame resistant reflective tape
  • Hook and loop closure for easy access
  • Non-magnetic; nickel- and ferrous-free
  • Lightweight and comfortable
  • Generous fit for wearer comfort
  • CComplies with RIS 3279-TOM for rail industry (orange only)
Certified to EN ISO 20471 Class 2 and EN ISO 14116 Index 1, the vest must be worn over index 2 or 3 garments to offer flame retardant protection.
Hi-Vis FR75 vests are available in yellow or orange.
Certifications: EN ISO 14116 Index 1. EN ISO 20471 Class 2. RIS 3279 TOM Issue 2 (orange only). CE certified.