Ultra Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Supplier: Brady
170772 170771 170773 170965
BRDY170772EA 84.72 EUR
BRDY170772 BRDY170771 BRDY170773 BRDY170965
Ultra Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Labels
Rough, glossy and irregular surfaces are no match for the ToughBond® ultra aggressive polyester label material (B-483). Print almost anywhere with a compact Brady handheld label maker. Compatible for BMP41, BMP51, BMP53, M511 Printers.
  • Versatile - Perfect for general identification and marking powder-coated and smooth plastic surfaces
  • Durable - Resistant to abrasions, extreme temperatures and solvents such as fuel, oil and water
  • Adhesive - Made with a permanent rubber-based adhesive that secures tightly to flat textured metals and smooth plastics alike
Label properties: Aggressive adhesive, heat resistant, low temperature.
Application: Asset tracking, barcode labeling, component and equipment labeling, data and telecommunications labeling, electrical labeling, general identification, inventory and inspection labeling.
Gloss finish, abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant B-483A labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Certifications: UL Recognized to UL969, CSA accepted, RoHS Compliant to 2011/65/EU (Pass 2015/863).