CRS 500 H₂O Filters
Supplier: Trajan Scientific and Medical
202270-B 202270-SS 202272-B 202272-SS
SGEA202270-BEA 178 EUR
SGEA202270-B SGEA202270-SS SGEA202272-B SGEA202272-SS
CRS 500 H₂O Filters
Filters Gas Filters
  • Capacity: 47 g
  • Efficiency: <5 ppb
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. flow rate 10 L per min
Nominal flow rate 2 L per min
Max. pressure 68,9 bar