Molecular weight markers, Amersham™ ECL™ Plex™ fluorescent Rainbow™ markers
Supplier: Cytiva
RPN851E RPN850E
RPN851EEA 351 EUR
RPN851E RPN850E
Molecular weight markers, Amersham™ ECL™ Plex™ fluorescent Rainbow™ markers
Electrophoresis Reagents Molecular Weight Markers
Amersham™ ECL™ Plex™ fluorescent Rainbow™ markers are protein molecular weight markers optimised for use with the ECL™ Plex™ Western blotting detection system. They are designed to provide visible marker bands on gels and membranes, as well as fluorescent images using Cy3 and Cy5 channels.
  • Broad colour spectrum for easier band identification
  • Sharper, more intense bands on gels and blots
  • Ready-to-load formulations