Western blotting combination pack, Amersham ECL Plex™
Supplier: Cytiva
RPN999
RPN998
RPN999EA
304
EUR
RPN999
RPN998
Western blotting combination pack, Amersham ECL Plex™
Electrophoresis Reagents
Western Blotting Reagents
Amersham ECL Plex™ Western blotting combination packs offer a complete system for fluorescent Western blotting, with improved formulation Cy3 conjugates for Lower background.
Delivery information: Each Combination Pack includes sufficient reagents for at least 1000 cm² of membrane: ECL Plex™ goat-α-mouse IgG-Cy3, 150 µg; ECL Plex™ goat-α-rabbit IgG-Cy5, 150 µg; ECL Plex™ fluorescent Rainbow™ markers, 120 µl; Hybond ECL™, 10×10 cm, 10 sheets or Hybond™-LFP, 20×20 cm, 3 sheets.