Safety spectacles, uvex skyguard NT
Supplier: UVEX
9175.261 9175.260
UVEX9175.261EA 26.8 EUR
UVEX9175.261 UVEX9175.260
Safety spectacles, uvex skyguard NT
Safety Glasses
The dual lens safety glasses were designed to provide an all-round protection. Based on the model Skylite spectacle a conceptually simple, injection moulded TPU frame was added.
  • TPU frame increases wearer protection levels, through minimising the risk of particle ingress
  • Flexible, closed-pore soft components of the frame enable an individual fit
  • Easy to clean
  • Adjustable length of side arms
EN 166-168, EN 170
Certifications: EN 166-168, EN 170