(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid 95%
Supplier: Apollo Scientific
Warning
OR80059 OR80059-100MG OR80059-1G
APOSOR80059EA 478 EUR
APOSOR80059 APOSOR80059-100MG APOSOR80059-1G
(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid 95%
(S)-3-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)propanoic acid
Formula: C₁₄H₁₈ClNO₄
MW: 299.75 g/mol
MDL Number: MFCD16394572
CAS Number: 1001180-04-6