Screen Quest™ Rhod-4 No Wash Calcium Assay Kit
Supplier: AAT BIOQUEST
36333 36334 36335
AATB36333EA 307 EUR
AATB36333 AATB36334 AATB36335
Screen Quest™ Rhod-4 No Wash Calcium Assay Kit
Assays Cellular Assays
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • >250 times fluorescence increase once bound to calcium
  • long-wavelength signal emission wavelength
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Brighter and more photoStable signal
Caution: For research use only.