2,3-Butanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,3-Dihydroxy butane, (±)-2,3-Butanediol, 2,3-Butylene glykol
M-Clarity™ quality level = MQ200
B84904-100G B84904-500G
SIALB84904-100GEA 183 EUR
SIALB84904-100G SIALB84904-500G
2,3-Butanediol, Sigma-Aldrich®
2,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 184
Melting Pt: 18…20 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004523
CAS Number: 513-85-9
EINECS: 208-173-6
Merck Index: 12,01602