ReadiLink™ iFluor® 555 Oligo and ssDNA Labelling Kit
Supplier: AAT BIOQUEST
17482 17483
AATB17482EA 353 EUR
AATB17482 AATB17483
ReadiLink™ iFluor® 555 Oligo and ssDNA Labelling Kit
Protein Linking and Labeling Systems
The kit is optimised for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
  • Robust protocol
  • Bright labeling
  • Photostable fluorescence
The resulting iFluor® 555-labelled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridisations..
Packaging: Packaged in plastic vials.
Caution: For research use only.