Buccutite™ Rapid PE Antibody Labelling Kits (2 Labellings)
Supplier: AAT BIOQUEST
1312 1310
AATB1312EA 460 EUR
AATB1312 AATB1310
Buccutite™ Rapid PE Antibody Labelling Kits (2 Labellings)
Protein Linking and Labeling Systems
R-Phycoerythrin (PE) is an orange fluorescent protein which has an excitation wavelength of 565 nm and an emission wavelength of 575 nm.
  • High conjugation yield
  • Stable crosslinking
  • Minimal hands-on time
  • Robust protocol
Order Now
SPECIFICATIONS
Environmentally Preferable
Host
Learn more