7-Amino-4-methylcoumarin ≥90%
Supplier: AAT BIOQUEST
Warning
Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC
51 53 52
AATB51EA 120 EUR
AATB51 AATB53 AATB52
7-Amino-4-methylcoumarin ≥90%
7-Amino-4-methylcoumarin
This compound is the precursor for a variety of AMC enzyme substrates.
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352
Melting Pt: 226
MDL Number: MFCD00006868
CAS Number: 26093-31-2
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7232a90dc4e2e8ed29c78554b52f5d0d649282ae&vid=aae157790e6d58d2607c526391a8df661c973e39&prid=web&clid=SAPDAM