Gibberellic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
M-Clarity™ quality level = MQ200
G7645-500MG G7645-10G G7645-1G G7645-5G
SIALG7645-500MGEA 77.4 EUR
SIALG7645-500MG SIALG7645-10G SIALG7645-1G SIALG7645-5G
Gibberellic acid, Sigma-Aldrich®
Gibberellic acid
Formula: C₁₉H₂₂O₆
Melting Pt: 234 °C
Density: 1.386 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079329
CAS Number: 77-06-5
EINECS: 201-001-0
Merck Index: 13,04430