2-[2-(Diethylamino)ethoxy]ethanol 98%
Supplier: Apollo Scientific
Danger
Synonyms: Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether, 2-(2-(Diethylamino)ethoxy)ethanol
OR019105-5G OR019105-25G OR019105-100G
APOSOR019105-5GEA 21.9 EUR
APOSOR019105-5G APOSOR019105-25G APOSOR019105-100G
2-[2-(Diethylamino)ethoxy]ethanol 98%
2-[2-(Diethylamino)ethoxy]ethanol
Formula: C₈H₁₉NO₂
MW: 161.25 g/mol
Boiling Pt: 95 °C
Density: 0.954
Flash Pt: 93 °C
MDL Number: MFCD00020604
CAS Number: 140-82-9