tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate, 1-Oxa-6-azaspiro[2.5]octane, N-BOC protected
OR01993-5G OR01993-25G
APOSOR01993-5GEA 72.5 EUR
APOSOR01993-5G APOSOR01993-25G
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
Melting Pt: 58…59 °C
MDL Number: MFCD07779385
CAS Number: 147804-30-6