2-Bromo-3,6-difluorobenzonitrile 95%
Supplier: Apollo Scientific
Warning
PC501831-250MG PC501831-1G
APOSPC501831-250MGEA 214 EUR
APOSPC501831-250MG APOSPC501831-1G
2-Bromo-3,6-difluorobenzonitrile 95%
2-Bromo-3,6-difluorobenzonitrile
Formula: C₇H₂BrF₂N
MW: 217.998 g/mol
Melting Pt: 31…32 °C
MDL Number: MFCD26687961
CAS Number: 1502090-29-0