2-(Aminomethyl)-5-chloro-3-fluoropyridine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: (5-Chloro-3-fluoropyridin-2-yl)methylamine hydrochloride, 5-Chloro-3-fluoropicolinylamine hydrochloride, (5-Chloro-3-fluoropyridin-2-yl)methanamine hydrochloride
PC10651-250MG PC10651-50MG PC10651-1G
APOSPC10651-250MGEA 352 EUR
APOSPC10651-250MG APOSPC10651-50MG APOSPC10651-1G
2-(Aminomethyl)-5-chloro-3-fluoropyridine hydrochloride
2-(Aminomethyl)-5-chloro-3-fluoropyridine hydrochloride
Formula: C₆H₆ClFN₂·ClH
MW: 197.038 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17214202
CAS Number: 1257535-29-7