But-3-yn-1-amine 96%
Supplier: Apollo Scientific
Danger
OR72152-250MG OR72152-1G OR72152-5G
APOSOR72152-250MGEA 71.2 EUR
APOSOR72152-250MG APOSOR72152-1G APOSOR72152-5G
But-3-yn-1-amine 96%
But-3-yn-1-amine
Formula: C₄H₇N
Boiling Pt: 100…103 °C
Density: 0.844
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09997741
CAS Number: 14044-63-4
UN: 2733
ADR: 3+8,II